siRNA Details
Gene Symbol: Rbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011254.5 | NP_035384.1 | 2 | 2193 | ||
GenBank mRNA | AK011641.1 | - | 2 | 341 | ||
BC018254.1 | - | 2 | 320 | 3498107 | ||
BC091751.1 | - | 2 | 337 | 4022443 | ||
X60367.1 | - | 2 | 2192 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
