siRNA Details
Gene Symbol: Rbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011254.5 | NP_035384.1 | 2 | 2181 | ||
GenBank mRNA | AK011641.1 | - | 2 | 329 | ||
BC018254.1 | - | 2 | 308 | 3498107 | ||
BC091751.1 | - | 2 | 325 | 4022443 | ||
X60367.1 | - | 2 | 2180 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_012733, M19257, FQ218090, FQ209927, FQ209770, and others.
Gene Ontology Categories:
