siRNA Details
Gene Symbol: Rdh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134006.4 | NP_598767.1 | 4, 5 | 915 | ||
GenBank mRNA | AF013288.1 | - | 4, 5 | 786 | ||
AF033195.1 | - | 4, 5 | 853 | |||
AF033196.2 | - | 4, 5 | 881 | |||
AK135139.1 | - | 4, 5 | 916 | |||
BC021372.1 | - | 4, 5 | 828 | 5040142 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
