siRNA Details
Gene Symbol: Rdx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104616.1 | NP_001098086.1 | 5 | 595 | ||
NM_001104617.1 | NP_001098087.1 | 5 | 1001 | |||
NM_009041.3 | NP_033067.2 | 5 | 1001 | |||
GenBank mRNA | AK016711.1 | - | 5 | 672 | ||
AK088292.1 | - | 5 | 708 | |||
AK146423.1 | - | 5 | 659 | |||
AK156370.1 | - | 5 | 687 | |||
AK162204.1 | - | 5 | 573 | |||
AK168449.1 | - | 5 | 672 | |||
BC002041.1 | - | 5 | 646 | |||
BC026786.1 | - | 5 | 674 | |||
BC034438.1 | - | 5 | 827 | |||
BC053417.1 | - | 5 | 675 | 6707873 | ||
DV662898.1 | - | 5 | 595 | |||
X60672.1 | - | 5 | 614 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
