siRNA Details
Gene Symbol: Rel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009044.2 | NP_033070.2 | 10 | 1566 | ||
GenBank mRNA | AK031627.1 | - | 10 | 1364 | ||
BC139447.1 | - | 10 | 1506 | 8862469 | ||
BC139448.1 | - | 10 | 1506 | 8862470 | ||
BC139770.1 | - | 10 | 1508 | 40111461 | ||
X15842.1 | - | 10 | 1688 | |||
X60271.1 | - | 10 | 1565 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
