siRNA Details
Gene Symbol: Rela | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009045.4 | NP_033071.1 | 11 | 1825 | ||
GenBank mRNA | AK089489.1 | - | 11 | 1570 | ||
AK144565.1 | - | 11 | 1572 | |||
AK154630.1 | - | 11 | 1570 | |||
AK155321.1 | - | 11 | 1834 | |||
AK155889.1 | - | 11 | 1567 | |||
AK157076.1 | - | 11 | 1557 | |||
AK158072.1 | - | 11 | 1835 | |||
AK212853.1 | - | 1 | 243 | |||
BC003818.1 | - | 11 | 1584 | 3489295 | ||
BC094053.1 | - | 11 | 1825 | 3157927 | ||
DQ020177.1 | - | 9 | 1394 | |||
M61909.1 | - | 11 | 1540 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_199267, BC079457, AY307375.
Gene Ontology Categories:
