siRNA Details
Gene Symbol: Dpf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291078.1 | NP_001278007.1 | 10, 11 | 1150 | ||
NM_011262.5 | NP_035392.1 | 9, 10 | 1108 | |||
GenBank mRNA | AK004812.1 | - | 9, 10 | 1045 | ||
AK138047.1 | - | 9, 10 | 1042 | |||
AK144106.1 | - | 9, 10 | 1047 | |||
AK144848.1 | - | 9, 10 | 1043 | |||
AK145851.1 | - | 9, 10 | 1104 | |||
AK156252.1 | - | 8, 9 | 821 | |||
U10435.1 | - | 9, 10 | 995 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
