siRNA Details
Gene Symbol: 2610305D13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145078.2 | NP_659544.2 | 5 | 944 | ||
GenBank mRNA | BC032186.1 | - | 1 | 432 | ||
BC058858.1 | - | 5 | 949 | |||
BC138179.1 | - | 2 | 587 | 8861198 | ||
BC145854.1 | - | 2 | 587 | 40131157 |
Gene Symbol: Gm13247 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243138.1 | NP_001230067.1 | 5 | 936 | ||
NM_001243139.1 | NP_001230068.1 | 4 | 821 |
Gene Symbol: Rex2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177767.1 | NP_001171238.1 | 5 | 935 | ||
GenBank mRNA | AF051348.1 | - | 948 | |||
BC067397.1 | - | 3 | 690 | 30462971 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
