siRNA Details
Gene Symbol: Trim27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009054.3 | NP_033080.2 | 8 | 1797 | ||
GenBank mRNA | AK031744.1 | - | 8 | 1515 | ||
AK088180.1 | - | 8 | 1281 | |||
AK207897.1 | - | 3 | 347 | |||
BC003219.1 | - | 8 | 1302 | 3482042 | ||
BC069924.1 | - | 8 | 1508 | |||
BC085503.1 | - | 8 | 1499 | |||
L46855.1 | - | 1408 | ||||
X75343.1 | - | 8 | 1212 |

More Information
Other Species Targeted

Gene Ontology Categories:
