siRNA Details
Gene Symbol: Rgs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009061.4 | NP_033087.2 | 4, 5 | 479 | ||
GenBank mRNA | AF215668.1 | - | 4, 5 | 464 | ||
AF432916.1 | - | 4, 5 | 458 | |||
AK029310.1 | - | 4, 5 | 481 | |||
AK031603.1 | - | 4, 5 | 481 | |||
AK077922.1 | - | 4, 5 | 480 | |||
AK162276.1 | - | 4, 5 | 481 | |||
BC023001.1 | - | 4, 5 | 462 | 5365271 | ||
U67187.1 | - | 4, 5 | 468 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
