siRNA Details
Gene Symbol: Rgs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009062.3 | NP_033088.2 | 4 | 431 | ||
GenBank mRNA | AB004315.1 | - | 4 | 312 | ||
AK030536.1 | - | 4 | 432 | |||
AK039134.1 | - | 4 | 432 | |||
BC003882.1 | - | 4 | 409 | 3496887 | ||
BC055293.1 | - | 4 | 421 | 6494449 | ||
DQ346659.1 | - | 4 | 431 | |||
DQ346660.1 | - | 4 | 431 | |||
DQ346661.1 | - | 4 | 431 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
