siRNA Details
Gene Symbol: Ring1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK155509.1 | - | 4 | 549 | ||
BC009070.1 | - | 4 | 536 | 2651194 | ||
CT010219.1 | - | 2 | 263 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
