siRNA Details
Gene Symbol: Xpr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011273.2 | NP_035403.1 | 13 | 1949 | ||
GenBank mRNA | AF114753.1 | - | 13 | 1834 | ||
AF131096.1 | - | 13 | 1739 | |||
AF198104.1 | - | 13 | 1739 | |||
AK048887.1 | - | 1 | 670 | |||
AK082228.1 | - | 1 | 1764 | |||
AK087140.1 | - | 4 | 1125 | |||
AK141660.1 | - | 13 | 1921 | |||
AK147403.1 | - | 13 | 1817 | |||
BC153872.1 | - | 13 | 1825 | 40110725 | ||
HQ323678.1 | - | 4 | 590 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
