siRNA Details
Gene Symbol: Rlim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011276.3 | NP_035406.3 | 5 | 1422 | ||
GenBank mRNA | AF069992.1 | - | 5 | 1377 | ||
AK013207.1 | - | 5 | 1394 | |||
AK029295.1 | - | 5 | 1416 | |||
BC012960.1 | - | 3 | 1155 | 4193003 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
