siRNA Details
Gene Symbol: Rlim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011276.3 | NP_035406.3 | 3 | 447 | ||
GenBank mRNA | AF069992.1 | - | 3 | 402 | ||
AK013207.1 | - | 3 | 419 | |||
AK029295.1 | - | 3 | 441 | |||
BC012960.1 | - | 1 | 180 | 4193003 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
