siRNA Details
Gene Symbol: Rnf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF169300.1 | - | 8 | 768 | ||
AK019171.1 | - | 8 | 783 | |||
AK019656.1 | - | 8 | 819 | |||
AK037465.1 | - | 183 | ||||
AK090162.1 | - | 8 | 806 | |||
AK145596.1 | - | 8 | 745 | |||
AK147057.1 | - | 8 | 728 | |||
AK151116.1 | - | 8 | 776 | |||
AK158987.1 | - | 8 | 785 | |||
AK159778.1 | - | 8 | 777 | |||
AK159949.1 | - | 8 | 825 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
