siRNA Details
Gene Symbol: Rnps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080127.1 | NP_001073596.1 | 3 | 292 | ||
NM_001080128.1 | NP_001073597.1 | 2 | 111 | |||
NM_009070.2 | NP_033096.2 | 4 | 414 | |||
GenBank mRNA | AK011475.1 | - | 3 | 294 | ||
AK146443.1 | - | 2 | 110 | |||
AK151150.1 | - | 3 | 282 | |||
AK165905.1 | - | 2 | 98 | |||
AK178555.1 | - | 2 | 243 | |||
AK182154.1 | - | 1 | 62 | |||
AK206201.1 | - | 2 | 248 | |||
AK211825.1 | - | 1 | 62 | |||
AK213636.1 | - | 1 | 62 | |||
BB868062.1 | - | 3 | 236 | |||
BC002061.1 | - | 3 | 280 | 3485311 | ||
BY013195.1 | - | 2 | 112 | |||
CJ058744.1 | - | 3 | 294 | |||
X70067.1 | - | 3 | 302 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
