siRNA Details
Gene Symbol: Rora | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289916.1 | NP_001276845.1 | 4 | 711 | ||
NM_013646.2 | NP_038674.1 | 5 | 720 | |||
GenBank mRNA | AK043990.1 | - | 1 | 80 | ||
AK149294.1 | - | 4 | 601 | |||
AK155827.1 | - | 4 | 662 | |||
BC003757.1 | - | 4 | 660 | 3592667 | ||
S82720.1 | - | 4 | 637 | |||
U53228.1 | - | 5 | 704 | |||
Y08640.1 | - | 4 | 866 | |||
Z82994.1 | - | 4 | 699 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
