siRNA Details
Gene Symbol: Rpgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177950.1 | NP_001171421.1 | 4, 5 | 518 | ||
NM_001177951.1 | NP_001171422.1 | 4, 5 | 518 | |||
NM_001177952.1 | NP_001171423.1 | 4, 5 | 512 | |||
NM_001177953.1 | NP_001171424.1 | 4, 5 | 518 | |||
NM_001177954.1 | NP_001171425.1 | 4, 5 | 518 | |||
NM_011285.2 | NP_035415.1 | 4, 5 | 518 | |||
GenBank mRNA | AF044677.1 | - | 4, 5 | 436 | ||
AJ238396.1 | - | 4, 5 | 430 | |||
AK017192.1 | - | 4, 5 | 450 | |||
AK029864.1 | - | 4, 5 | 644 | |||
AK038652.1 | - | 4, 5 | 482 | |||
AK081631.1 | - | 4, 5 | 457 | |||
AK136298.1 | - | 4, 5 | 449 | |||
AK138977.1 | - | 4, 5 | 513 |
