siRNA Details
Gene Symbol: Mrpl23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011288.1 | NP_035418.1 | 3 | 222 | ||
GenBank mRNA | AB049646.1 | - | 3 | 176 | ||
AK086805.1 | - | 3 | 176 | |||
BC038626.1 | - | 3 | 194 | 4008503 | ||
BC081460.1 | - | 3 | 162 | 6814073 | ||
U84902.1 | - | 3 | 164 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
