siRNA Details
Gene Symbol: Rpl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016980.2 | NP_058676.1 | 2, 3 | 199 | ||
GenBank mRNA | AK008489.1 | - | 2, 3 | 160 | ||
AK013107.1 | - | 2, 3 | 157 | |||
AK032778.1 | - | 2, 3 | 156 | |||
AK088036.1 | - | 2, 3 | 159 | |||
AK152381.1 | - | 2, 3 | 156 | |||
AK166865.1 | - | 2, 3 | 118 | |||
AK169313.1 | - | 2, 3 | 157 | |||
BC005790.1 | - | 2, 3 | 139 | |||
BC026934.1 | - | 2, 3 | 137 | 2651564 | ||
BC083318.1 | - | 2, 3 | 139 | 6818756 | ||
BC091752.1 | - | 2, 3 | 137 | 4208431 | ||
BC106128.1 | - | 2, 3 | 107 | 2651407 | ||
CJ073701.1 | - | 2, 3 | 199 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
