siRNA Details
Gene Symbol: Rpl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011292.2 | NP_035422.1 | 8 | 679 | ||
GenBank mRNA | AF260271.1 | - | 7 | 649 | ||
AK012445.1 | - | 8 | 649 | |||
AK017388.1 | - | 8 | 649 | |||
AK017427.1 | - | 8 | 649 | |||
AK050296.1 | - | 2 | 2607 | |||
AK053431.1 | - | 3 | 2646 | |||
AK084278.1 | - | 8 | 650 | |||
AK088166.1 | - | 8 | 649 | |||
AK153562.1 | - | 8 | 649 | |||
AK159980.1 | - | 8 | 649 | |||
AK168326.1 | - | 8 | 647 | |||
AK169217.1 | - | 8 | 647 | |||
AK191132.1 | - | 4 | 319 | |||
AK204218.1 | - | 4 | 252 | |||
BC013165.1 | - | 7 | 639 | 2655358 | ||
BC069963.1 | - | 1 | 1445 | |||
BC081435.1 | - | 7 | 625 | 6824107 | ||
BC083166.1 | - | 7 | 630 | 6397429 | ||
BC083329.1 | - | 7 | 628 | 6824346 | ||
BC086937.1 | - | 8 | 646 | 6787671 | ||
BC089319.1 | - | 7 | 628 | 6823996 | ||
BC093526.1 | - | 7 | 628 | 5150493 |
Gene Symbol: Gm12795 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AV373702.1 | - | 1 | 196 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
