siRNA Details
Gene Symbol: Sub1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011294.3 | NP_035424.1 | 4 | 322 | ||
GenBank mRNA | AK030954.1 | - | 1 | 1273 | ||
AK048371.1 | - | 4 | 322 | |||
AK049260.1 | - | 4 | 322 | |||
AK132021.1 | - | 4 | 321 | |||
AK146335.1 | - | 4 | 321 | |||
AK151734.1 | - | 4 | 322 | |||
AK154114.1 | - | 4 | 321 | |||
BC010967.1 | - | 4 | 320 | 4018261 | ||
J03750.1 | - | 4 | 306 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
