siRNA Details
Gene Symbol: Rps12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011295.6 | NP_035425.2 | 5 | 381 | ||
GenBank mRNA | AK002848.1 | - | 5 | 373 | ||
AK008057.1 | - | 5 | 373 | |||
AK032757.1 | - | 5 | 373 | |||
AK132208.1 | - | 3 | 1357 | |||
AK166923.1 | - | 5 | 371 | |||
AK166992.1 | - | 5 | 376 | |||
AK168324.1 | - | 5 | 373 | |||
AK169215.1 | - | 5 | 373 | |||
AK210130.1 | - | 3 | 194 | |||
BC002079.1 | - | 5 | 357 | 3489757 | ||
BC089338.1 | - | 5 | 369 | 6407567 | ||
BC089339.1 | - | 5 | 355 | 6408026 | ||
BC090257.1 | - | 5 | 348 | 4209024 | ||
BC092044.1 | - | 5 | 355 | 2655660 | ||
BC096653.1 | - | 5 | 384 | 1528457 | ||
BC099377.1 | - | 5 | 357 | 30936226 | ||
CJ207519.1 | - | 3 | 260 | |||
X15962.1 | - | 4 | 346 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
