siRNA Details
Gene Symbol: Rps16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013647.2 | NP_038675.2 | 4 | 389 | ||
GenBank mRNA | AK002904.1 | - | 1 | 327 | ||
AK010613.1 | - | 1 | 330 | |||
AK010641.1 | - | 1 | 330 | |||
AK011060.1 | - | 1 | 341 | |||
AK019400.2 | - | 1 | 328 | |||
AK083946.1 | - | 1 | 328 | |||
AK088413.1 | - | 1 | 329 | |||
AK088716.1 | - | 1 | 329 | |||
AK150615.1 | - | 1 | 320 | |||
AK150808.1 | - | 1 | 331 | |||
AK168207.1 | - | 1 | 328 | |||
AK169328.1 | - | 1 | 329 | |||
BC082286.1 | - | 1 | 308 | |||
BC090618.1 | - | 1 | 356 | 5713263 | ||
BC115673.1 | - | 1 | 296 | 40104131 | ||
BC115674.1 | - | 1 | 296 | 40104132 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
