siRNA Details
Gene Symbol: Rps24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011297.2 | NP_035427.2 | 6 | 741 | ||
NM_207634.1 | NP_997517.1 | 5 | 721 | |||
NM_207635.1 | NP_997518.1 | 7 | 759 | |||
GenBank mRNA | AK010856.1 | - | 5 | 685 | ||
AK049387.1 | - | 681 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
