siRNA Details
Gene Symbol: Rps4x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009094.1 | NP_033120.1 | 3 | 286 | ||
GenBank mRNA | AK002445.1 | - | 4 | 304 | ||
AK010624.1 | - | 4 | 307 | |||
AK010676.1 | - | 4 | 304 | |||
AK010921.1 | - | 4 | 304 | |||
AK012429.1 | - | 4 | 304 | |||
AK014210.1 | - | 4 | 302 | |||
AK088410.1 | - | 4 | 304 | |||
AK135369.1 | - | 4 | 307 | |||
AK153211.1 | - | 4 | 298 | |||
BC009100.1 | - | 3 | 311 | 3482299 | ||
BC089348.1 | - | 3 | 288 | |||
BC089349.1 | - | 3 | 282 | |||
BC106152.1 | - | 3 | 287 | 3669367 | ||
M73436.1 | - | 3 | 286 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
