siRNA Details
Gene Symbol: Ryr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009109.2 | NP_033135.2 | 98, 99 | 14438 | ||
GenBank mRNA | AY268935.1 | - | 97, 98 | 14438 | ||
BC025202.1 | - | 4, 5 | 575 | |||
BC051248.1 | - | 6, 7 | 807 | |||
BC055487.1 | - | 5, 6 | 588 | |||
X83932.1 | - | 6, 7 | 696 |
