siRNA Details
Gene Symbol: S100a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011309.3 | NP_035439.1 | 3 | 311 | ||
GenBank mRNA | AF087687.1 | - | 3 | 278 | ||
AK002721.1 | - | 3 | 252 | |||
AK012578.1 | - | 3 | 311 | |||
AK168822.1 | - | 3 | 247 | |||
BC005590.1 | - | 3 | 383 | 3964547 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
