siRNA Details
Gene Symbol: S100a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016740.3 | NP_058020.1 | 2 | 149 | ||
GenBank mRNA | AK002490.1 | - | 2 | 151 | ||
AK003889.1 | - | 2 | 149 | |||
AK164352.1 | - | 2 | 150 | |||
BC021916.1 | - | 2 | 128 | 4913795 | ||
BC086903.1 | - | 2 | 148 | 6764162 | ||
U41341.1 | - | 1 | 83 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
