siRNA Details
Gene Symbol: S100a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011312.2 | NP_035442.1 | 1 | 109 | ||
GenBank mRNA | AF087469.1 | - | 1 | 109 | ||
AI327447.1 | - | 1 | 109 | |||
BC117108.1 | - | 1 | 108 | 40126427 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
