siRNA Details
Gene Symbol: S100a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011312.2 | NP_035442.1 | 1 | 80 | ||
GenBank mRNA | AF087469.1 | - | 1 | 80 | ||
AI327447.1 | - | 1 | 80 | |||
BC117108.1 | - | 1 | 79 | 40126427 |

More Information
Other Species Targeted

Gene Ontology Categories:
