siRNA Details
Gene Symbol: S100a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011313.2 | NP_035443.1 | 3 | 535 | ||
GenBank mRNA | AF549497.1 | - | 3 | 243 | ||
AK008241.1 | - | 3 | 246 | |||
AK008941.1 | - | 3 | 246 | |||
AK150408.1 | - | 3 | 246 | |||
AK151144.1 | - | 3 | 246 | |||
AK151587.1 | - | 3 | 246 | |||
BC003832.1 | - | 3 | 298 | 3490662 | ||
BC010774.1 | - | 3 | 260 | 4219642 | ||
BM932500.1 | - | 3 | 547 | |||
M37761.1 | - | 3 | 244 | |||
X52278.1 | - | 3 | 253 | |||
X66449.1 | - | 337 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
