siRNA Details
Gene Symbol: S100a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013650.2 | NP_038678.1 | 3 | 229 | ||
GenBank mRNA | AK157837.1 | - | 3 | 230 | ||
AV007095.1 | - | 2 | 172 | |||
BC078629.1 | - | 3 | 215 | 864657 | ||
M83218.1 | - | 2 | 191 | |||
S57123.1 | - | 3 | 216 | |||
Y09039.1 | - | 2 | 179 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
