siRNA Details
Gene Symbol: Saa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009117.3 | NP_033143.1 | 3 | 151 | ||
GenBank mRNA | AK136475.1 | - | 3 | 157 | ||
BC052495.1 | - | 1 | 1580 | |||
BC087933.1 | - | 3 | 151 | 30310171 |
Gene Symbol: Saa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011314.2 | NP_035444.1 | 3 | 154 | ||
GenBank mRNA | BC024606.1 | - | 2 | 128 | 3662957 | |
BC034858.1 | - | 1 | 1314 | |||
L22190.1 | - | 2 | 152 | |||
M11130.1 | - | 3 | 154 | |||
U60437.1 | - | 2 | 118 | |||
U60438.1 | - | 2 | 118 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
