siRNA Details
Gene Symbol: Acsm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016870.3 | NP_058566.3 | 5 | 768 | ||
NM_212441.2 | NP_997606.2 | 4 | 663 | |||
NM_212442.2 | NP_997607.2 | 4 | 566 | |||
GenBank mRNA | AB022340.2 | - | 4 | 554 | ||
AF068246.1 | - | 4 | 553 | |||
AK041060.1 | - | 4 | 538 | |||
AK077247.1 | - | 4 | 625 | |||
AK143946.1 | - | 566 | ||||
AK165516.1 | - | 4 | 540 | |||
AY064696.1 | - | 4 | 639 | |||
BC015248.1 | - | 5 | 728 | 4239288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
