siRNA Details
Gene Symbol: Satb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163630.1 | NP_001157102.1 | 4, 5 | 847 | ||
NM_001163631.1 | NP_001157103.1 | 4, 5 | 715 | |||
NM_001163632.1 | NP_001157104.1 | 4, 5 | 688 | |||
NM_009122.2 | NP_033148.2 | 4, 5 | 738 | |||
GenBank mRNA | AK037740.1 | - | 4, 5 | 738 | ||
AK088563.1 | - | 4, 5 | 714 | |||
AK134040.1 | - | 4, 5 | 718 | |||
AK158518.1 | - | 4, 5 | 848 | |||
BC011132.1 | - | 4, 5 | 598 | 4164993 | ||
CT010371.1 | - | 3, 4 | 526 | |||
U05252.1 | - | 4, 5 | 724 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
