siRNA Details
Gene Symbol: Satb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163630.1 | NP_001157102.1 | 7 | 1089 | ||
NM_001163631.1 | NP_001157103.1 | 7 | 957 | |||
NM_001163632.1 | NP_001157104.1 | 7 | 930 | |||
NM_009122.2 | NP_033148.2 | 7 | 980 | |||
GenBank mRNA | AK037740.1 | - | 7 | 980 | ||
AK088563.1 | - | 7 | 956 | |||
AK134040.1 | - | 7 | 960 | |||
AK158518.1 | - | 7 | 1090 | |||
BC011132.1 | - | 7 | 840 | 4164993 | ||
CT010371.1 | - | 6 | 768 | |||
U05252.1 | - | 7 | 966 |
