siRNA Details
Gene Symbol: Nkx1-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009123.2 | NP_033149.1 | 2 | 1471 | ||
GenBank mRNA | AF222444.1 | - | 2 | 1471 | ||
BC139757.1 | - | 2 | 1400 | 40047346 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
