siRNA Details
Gene Symbol: Serpinb3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009126.3 | NP_033152.3 | 6 | 675 | ||
GenBank mRNA | AY144683.1 | - | 6 | 659 | ||
AY144684.1 | - | 6 | 659 | |||
BC063756.1 | - | 6 | 675 | 30288465 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
