siRNA Details
Gene Symbol: Zc3h7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081016.1 | NP_001074485.1 | 12 | 1733 | ||
GenBank mRNA | AK129268.1 | - | 12 | 1410 | ||
AK147484.1 | - | 12 | 1497 | |||
AK147978.1 | - | 12 | 1506 | |||
AK157711.1 | - | 4 | 1103 | |||
AK165492.1 | - | 12 | 1525 | |||
AK171949.1 | - | 12 | 1448 | |||
AK193237.1 | - | 1 | 27 | |||
BC059923.1 | - | 12 | 1488 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
