siRNA Details
Gene Symbol: Msr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113326.1 | NP_001106797.1 | 4 | 576 | ||
NM_031195.2 | NP_112472.2 | 4 | 576 | |||
GenBank mRNA | AF203781.1 | - | 3 | 518 | ||
AK028480.1 | - | 4 | 570 | |||
AK089178.1 | - | 4 | 570 | |||
AK150518.1 | - | 4 | 577 | |||
AK151403.1 | - | 4 | 550 | |||
AK151855.1 | - | 4 | 577 | |||
AK154806.1 | - | 4 | 553 | |||
AK155362.1 | - | 4 | 572 | |||
AK155589.1 | - | 4 | 570 | |||
BC003814.1 | - | 4 | 547 | 3594142 | ||
D13382.1 | - | 3 | 518 | |||
L04274.1 | - | 4 | 523 | |||
L04275.1 | - | 4 | 523 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
