siRNA Details
Gene Symbol: Ccl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011329.3 | NP_035459.1 | 2 | 171 | ||
GenBank mRNA | AF065924.1 | - | 2 | 150 | ||
AF065925.1 | - | 2 | 150 | |||
AF065926.1 | - | 2 | 150 | |||
AK019040.1 | - | 2 | 171 | |||
BC042744.1 | - | 1782 | ||||
BC120806.1 | - | 2 | 162 | 40129729 | ||
M17957.1 | - | 2 | 174 | |||
M23501.1 | - | 2 | 171 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
