siRNA Details
Gene Symbol: Ccl17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011332.3 | NP_035462.2 | 3 | 437 | ||
GenBank mRNA | AF125570.1 | - | 3 | 526 | ||
AF125571.1 | - | 3 | 468 | |||
AF125572.1 | - | 3 | 441 | |||
AJ242587.1 | - | 3 | 416 | |||
AK007663.1 | - | 3 | 438 | |||
BC028505.1 | - | 3 | 421 | 3328278 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
