siRNA Details
Gene Symbol: Ccl21b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011335.2 | NP_035465.2 | 496 |
Gene Symbol: Ccl21a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011124.4 | NP_035254.1 | 4 | 533 | ||
GenBank mRNA | AF001980.1 | - | 4 | 460 | ||
AF006637.1 | - | 4 | 556 | |||
AK144258.1 | - | 4 | 535 | |||
BC025974.1 | - | 4 | 532 | 4208773 | ||
BC038120.1 | - | 4 | 502 | 3372243 |
Gene Symbol: Ccl21c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023052.2 | NP_075539.1 | 4 | 532 | ||
GenBank mRNA | BC087957.1 | - | 4 | 532 | 6750979 | |
U88322.1 | - | 4 | 505 |
Gene Symbol: Gm10591 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193668.1 | NP_001180597.1 | 532 |
Gene Symbol: Gm13304 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193666.1 | NP_001180595.1 | 532 |
Gene Symbol: Gm1987 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193667.1 | NP_001180596.1 | 4 | 535 |
Gene Symbol: Gm21541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270360.1 | NP_001257289.1 | 4 | 532 | ||
GenBank mRNA | AA123440.1 | - | 1 | 66 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
