siRNA Details
Gene Symbol: Ccl27a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048179.1 | NP_001041644.1 | 6 | 487 | ||
NM_011336.1 | NP_035466.1 | 3 | 321 | |||
GenBank mRNA | AB013398.1 | - | 3 | 321 | ||
AF082392.1 | - | 3 | 299 | |||
AF099931.1 | - | 3 | 309 | |||
AK005398.1 | - | 4 | 461 | |||
AK146066.1 | - | 3 | 1825 | |||
AY744154.1 | - | 1 | 629 | |||
AY744155.1 | - | 2 | 440 | |||
BC107226.2 | - | 3 | 321 | 40051915 | ||
CA451425.1 | - | 6 | 487 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
