siRNA Details
Gene Symbol: Ccl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009139.3 | NP_033165.1 | 2 | 238 | ||
GenBank mRNA | AF128188.1 | - | 2 | 134 | ||
AF128189.1 | - | 2 | 134 | |||
AF128190.1 | - | 2 | 134 | |||
AF128191.1 | - | 2 | 134 | |||
AF128192.1 | - | 2 | 134 | |||
AK002697.1 | - | 2 | 141 | |||
AK008547.1 | - | 2 | 142 | |||
AK150336.1 | - | 2 | 141 | |||
AK154553.1 | - | 2 | 142 | |||
BC002073.1 | - | 2 | 131 | 3492808 | ||
BY172106.1 | - | 2 | 238 | |||
M58004.1 | - | 2 | 141 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
