siRNA Details
Gene Symbol: Cxcl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009140.2 | NP_033166.1 | 3 | 264 | ||
GenBank mRNA | AK137628.1 | - | 3 | 292 | ||
AK150450.1 | - | 3 | 172 | |||
AK155458.1 | - | 1 | 500 | |||
AK155874.1 | - | 3 | 293 | |||
AK155916.1 | - | 3 | 291 | |||
AK157079.1 | - | 3 | 291 | |||
BC119511.1 | - | 3 | 238 | 40055368 | ||
BC119512.1 | - | 3 | 238 | 40055369 | ||
X53798.1 | - | 3 | 264 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
