siRNA Details
Gene Symbol: Sec61g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109971.1 | NP_001103441.1 | 4 | 730 | ||
NM_001109972.1 | NP_001103442.1 | 4 | 709 | |||
NM_011343.3 | NP_035473.2 | 4 | 705 | |||
GenBank mRNA | AK012393.1 | - | 4 | 348 | ||
AK012644.1 | - | 4 | 369 | |||
BC019129.1 | - | 4 | 383 | |||
BC019158.1 | - | 4 | 383 | 3487721 | ||
BC081456.1 | - | 3 | 333 | 6407982 | ||
BY468217.1 | - | 4 | 358 | |||
M88756.1 | - | 2 | 170 | |||
U11027.1 | - | 3 | 321 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
