siRNA Details
Gene Symbol: Sema5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013661.2 | NP_038689.2 | 10 | 1576 | ||
GenBank mRNA | BC052397.1 | - | 10 | 1576 | 5719939 | |
X97818.1 | - | 10 | 1367 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
